missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse CXCL16 Protein

Product Code. 18409981 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18409981

Brand: Novus Biologicals™ NBP2351661mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

CXCL16 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using murine lymphocytes is in a concentration of 20-1000 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8BSU2, Q8BSU2, Q8BSU2
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS.
Gene ID (Entrez) 58191
Molecular Weight (g/mol) M.W. (Theoretical): 9.9 kDa
Quantity 1mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of CXCL16 as determined by LAL method.
Gene Alias chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Gene Symbol CXCL16
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Research Category Biologically Active Proteins, Chemokines and Cytokines
Purity or Quality Grade >98% pure by SDS-PAGE and HPLC
Protein CXCL16
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.