missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse CXCL4/PF4 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
PF4/Platelet factor 4 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human neutrophils is in a concentration of 10-100ng/mL.
Specifications
Specifications
| Accession Number | Q9Z126, Q9Z126, Q9Z126 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, 1.5 M NaCl, pH 7.4. |
| Gene ID (Entrez) | 5196 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 8.2 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of CXCL4/PF4 as determined by LAL method. |
| Gene Alias | chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, PF-4/CXCL4, platelet factor 4, SCYB4oncostatin-A |
| Gene Symbol | PF4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction