missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse CXCL4/PF4 Protein

Product Code. 18488461 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18488461

Brand: Novus Biologicals™ NBP235119100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

PF4/Platelet factor 4 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human neutrophils is in a concentration of 10-100ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z126, Q9Z126, Q9Z126
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, 1.5 M NaCl, pH 7.4.
Gene ID (Entrez) 5196
Molecular Weight (g/mol) M.W. (Theoretical): 8.2 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of CXCL4/PF4 as determined by LAL method.
Gene Alias chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, PF-4/CXCL4, platelet factor 4, SCYB4oncostatin-A
Gene Symbol PF4
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Research Category Biologically Active Proteins, Cancer, Signal Transduction
Purity or Quality Grade >97%pure by SDS-PAGE and HPLC
Protein CXCL4/PF4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.