missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Endoplasmic reticulum aminopeptidase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Endoplasmic reticulum aminopeptidase 1, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_109636.1 |
---|---|
Gene ID (Entrez) | 80898 |
Name | Endoplasmic reticulum aminopeptidase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939199
|
enQuireBio™
QP5992-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959199
|
enQuireBio™
QP5992-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949199
|
enQuireBio™
QP5992-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969199
|
enQuireBio™
QP5992-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979199
|
enQuireBio™
QP5992-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15989199
|
enQuireBio™
QP5992-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_109636.1 | |
Endoplasmic reticulum aminopeptidase 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
80898 | |
Research Use Only | |
Erap1 | |
Recombinant Protein | |
E. coli | |
PCVQRAERYF REWKSSNGNM SIPIDVTLAV FAVGAQNTEG WDFLYSKYQS SLSSTEKSQI EFSLCTSKDP EKLQWLLDQS FKGEIIKTQE FPHILTLIGR NPVGYPLAWK FLRENWNKLV QKFELGSSSI AHMVMGTTDQ FSTRARLEEV KGFFSSLKEN GSQLRCVQQT IETIEENIRW MDKNFDKIRL WLQKEKPELL | |
Greater than 80.6% as determined by SDS-PAGE. |