missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse FOLH1 Protein

A cDNA sequence encoding the FOLH1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11895-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958718

  • 203.00 € / 10µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

53320
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
ADPKPSNEAT GNVSHSGMKK EFLHELKAEN IKKFLYNFTR TPHLAGTQNN FELAKQIHDQ WKEFGLDLVE LSHYDVLLSY PNKTHPNYIS IINEDGNEIF KTSLSEQPPP GYENISDVVP PYSAFSPQGT PEGDLVYVNY ARTEDFFKLE REMKISCSGK IVIARYGKVF RGNMVKNAQL AGAKGMILYS DPADYFVPAV KSYPDGWNLP GGGVQRGNVL NLNGAGDPLT PGYPANEHAY RHELTNAVGL PSIPVHPIGY DDAQKLLEHM GGPAPPDSSW KGGLKVPYNV GPGFAGNFST QKVKMHIHSY TKVTRIYNVI GTLKGALEPD RYVILGGHRD AWVFGGIDPQ SGAAVVHEIV RSFGTLKKKG RRPRRTILFA SWDAEEFGLL GSTEWAEEHS RLLQERGVAY INADSSIEGN YTLRVDCTPL MYSLVYNLTK ELQSPDEGFE GKSLYDSWKE KSPSPEFIGM PRISKLGSGN DFEVFFQRLG IASGRARYTK NWKTNKVSSY PLYHSVYETY ELVVKFYDPT FKYHLTVAQV RGAMVFELAN SIVLPFDCQS YAVALKKYAD TIYNISMKHP QEMKAYMISF DSLFSAVNNF TDVASKFNQR LQELDKSNPI LLRIMNDQLM YLERAFIDPL GLPGRPFYRH IIYAPSSHNK YAGESFPGIY DALFDISSKV NASKAWNEVK RQISIATFTV QAAAETLREV AHHHHHH
Greater than 85% as determined by SDS-PAGE.
FOLH1 Protein
Research Use Only
Folh1
Mouse
His
The FOLH1 solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers