missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse FOLH1 Protein
A cDNA sequence encoding the FOLH1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 53320 |
---|---|
Name | FOLH1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Folh1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978718
|
enQuireBio™
QP11895-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958718
|
enQuireBio™
QP11895-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968718
|
enQuireBio™
QP11895-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
53320 | |
Research Use Only | |
Folh1 | |
Mouse | |
His | |
The FOLH1 solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
FOLH1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPKPSNEAT GNVSHSGMKK EFLHELKAEN IKKFLYNFTR TPHLAGTQNN FELAKQIHDQ WKEFGLDLVE LSHYDVLLSY PNKTHPNYIS IINEDGNEIF KTSLSEQPPP GYENISDVVP PYSAFSPQGT PEGDLVYVNY ARTEDFFKLE REMKISCSGK IVIARYGKVF RGNMVKNAQL AGAKGMILYS DPADYFVPAV KSYPDGWNLP GGGVQRGNVL NLNGAGDPLT PGYPANEHAY RHELTNAVGL PSIPVHPIGY DDAQKLLEHM GGPAPPDSSW KGGLKVPYNV GPGFAGNFST QKVKMHIHSY TKVTRIYNVI GTLKGALEPD RYVILGGHRD AWVFGGIDPQ SGAAVVHEIV RSFGTLKKKG RRPRRTILFA SWDAEEFGLL GSTEWAEEHS RLLQERGVAY INADSSIEGN YTLRVDCTPL MYSLVYNLTK ELQSPDEGFE GKSLYDSWKE KSPSPEFIGM PRISKLGSGN DFEVFFQRLG IASGRARYTK NWKTNKVSSY PLYHSVYETY ELVVKFYDPT FKYHLTVAQV RGAMVFELAN SIVLPFDCQS YAVALKKYAD TIYNISMKHP QEMKAYMISF DSLFSAVNNF TDVASKFNQR LQELDKSNPI LLRIMNDQLM YLERAFIDPL GLPGRPFYRH IIYAPSSHNK YAGESFPGIY DALFDISSKV NASKAWNEVK RQISIATFTV QAAAETLREV AHHHHHH | |
Greater than 85% as determined by SDS-PAGE. |