missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GAD65 / GAD2 Protein
A DNA sequence encoding the Mus musculus (Mouse) GAD65 / GAD2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6062-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_032104.2 | |
GAD65 / GAD2 Protein | |
Research Use Only | |
Gad2 | |
Recombinant Protein | |
E. coli | |
MASPGSGFWS FGSEDGSADP ENPGTARAWC QVAQKFTGGI GNKLCALLYG DSGKPAEGGG SVTSRAATGK VACTCDQKPC NCPKGDVNYA FLHATDLLPA CDGERPTLAF LQDVMNILLQ YVVKSFDRST KVIDFHYPNE LLQEYNWELA DQPQNLEEIL THCQTTLKYA IKTGHPRYFN QLSTGLDMVG LAADWLTSTA NTNMFTYEIA PVFVLLEYVT LKKMREIIGW PGGSGDGIFS PGGAISNMYA MLIARYKMFP EVKEKGMAAV PRLIAFTSEH SHFSLKKGAA ALGIGTDSVI LIKCDERGKM IPSDLERRIL EVKQKGFVPF LVSATAGTTV YGAFDPLLAV ADICKKYKIW MHVDAAWGGG LLMSRKHKWK LSGVERANSV TWNPHKMMGV PLQCSALLVR EEGLMQSCNQ MHASYLFQQD KHYDLSYDTG DKALQCGRHV DVFKLWLMWR AKGTTGFEAH IDKCLELAEY LYTIIKNREG YEMVFDGKPQ HTNVCFWFVP PSLRTLEDNE ERMSRLSKVA PVIKARMMEY GTTMVSYQPL GDKVNFFRMV ISNPAATHQD IDFLIEEIER LGQDL | |
Greater than 90% as determined by SDS-PAGE. |
14417 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |