missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse HPGD Protein

A cDNA sequence encoding the HPGD was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12300-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15909988

  • 131.00 € / 5µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

15446
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMHVNGKV ALVTGAAQGI GKAFAEALLL HGAKVALVDW NLEAGVKCKA ALDEQFEPQK TLFVQCDVAD QKQLRDTFRK VVDHFGRLDI LVNNAGVNNE KNWEQTLQIN LVSVISGTYL GLDYMSKQNG GEGGIIINMS SLAGLMPVAQ QPVYCASKHG IIGFTRSAAM AANLMKSGVR LNVICPGFVD TPILESIEKE ENMGQYIEYK DQIKAMMKFY GVLHPSTIAN GLINLIEDDA LNGAIMKITA SKGIHFQDYD ISPLLVKAPL TS
Greater than 95.0% as determined by SDS-PAGE.
HPGD Protein
Research Use Only
Hpgd
Mouse
His
The HPGD solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers