missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HPGD Protein
A cDNA sequence encoding the HPGD was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 15446 |
---|---|
Name | HPGD Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Hpgd |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909988
|
enQuireBio™
QP12300-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999978
|
enQuireBio™
QP12300-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989978
|
enQuireBio™
QP12300-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
15446 | |
Research Use Only | |
Hpgd | |
Mouse | |
His | |
The HPGD solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
HPGD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMHVNGKV ALVTGAAQGI GKAFAEALLL HGAKVALVDW NLEAGVKCKA ALDEQFEPQK TLFVQCDVAD QKQLRDTFRK VVDHFGRLDI LVNNAGVNNE KNWEQTLQIN LVSVISGTYL GLDYMSKQNG GEGGIIINMS SLAGLMPVAQ QPVYCASKHG IIGFTRSAAM AANLMKSGVR LNVICPGFVD TPILESIEKE ENMGQYIEYK DQIKAMMKFY GVLHPSTIAN GLINLIEDDA LNGAIMKITA SKGIHFQDYD ISPLLVKAPL TS | |
Greater than 95.0% as determined by SDS-PAGE. |