missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HPGD Protein
A cDNA sequence encoding the HPGD was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12300-20ug
Additional Details : Weight : 0.01000kg
Specifications
15446 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMHVNGKV ALVTGAAQGI GKAFAEALLL HGAKVALVDW NLEAGVKCKA ALDEQFEPQK TLFVQCDVAD QKQLRDTFRK VVDHFGRLDI LVNNAGVNNE KNWEQTLQIN LVSVISGTYL GLDYMSKQNG GEGGIIINMS SLAGLMPVAQ QPVYCASKHG IIGFTRSAAM AANLMKSGVR LNVICPGFVD TPILESIEKE ENMGQYIEYK DQIKAMMKFY GVLHPSTIAN GLINLIEDDA LNGAIMKITA SKGIHFQDYD ISPLLVKAPL TS | |
Greater than 95.0% as determined by SDS-PAGE. |
HPGD Protein | |
Research Use Only | |
Hpgd | |
Mouse | |
His | |
The HPGD solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |