missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IFNG / Interferon Gamma Protein
Recombinant Protein
Brand: enQuireBio™ QP4141-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032363.1 | |
IFNG / Interferon Gamma Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
MHGTVIESLE SLNNYFNSSG IDVEEKSLFL DIWRNWQKDG DMKILQSQII SFYLRLFEVL KDNQAISNNI SVIESHLITT FFSNSKAKKD AFMSIAKFEV NNPQVQRQAF NELIRVVHQL LPESSLRKRK RSRC | |
Greater than 95% as determined by SDS-PAGE. |
15978 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the Viral CPE assay using EMC virus on L929 cells assay. The bioactivity acceptance criteria is set at 1 x 10^7 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5 |