missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IFNG / Interferon Gamma Protein
Recombinant Protein
107.00 € - 1595.00 €
Specifications
Accession Number | NP_032363.1 |
---|---|
Gene ID (Entrez) | 15978 |
Name | IFNG / Interferon Gamma Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975579
|
enQuireBio™
QP4141-20UG |
20 μg |
107.00 €
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985579
|
enQuireBio™
QP4141-250UG |
250 μg |
555.00 €
250µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995579
|
enQuireBio™
QP4141-500UG |
500 μg |
940.00 €
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965579
|
enQuireBio™
QP4141-1MG |
1 mg |
1595.00 €
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955579
|
enQuireBio™
QP4141-100 UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032363.1 | |
IFNG / Interferon Gamma Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the Viral CPE assay using EMC virus on L929 cells assay. The bioactivity acceptance criteria is set at 1 x 10^7 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5 |
15978 | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
MHGTVIESLE SLNNYFNSSG IDVEEKSLFL DIWRNWQKDG DMKILQSQII SFYLRLFEVL KDNQAISNNI SVIESHLITT FFSNSKAKKD AFMSIAKFEV NNPQVQRQAF NELIRVVHQL LPESSLRKRK RSRC | |
Greater than 95% as determined by SDS-PAGE. |