missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-15 Protein
Recombinant Protein
Brand: enQuireBio™ QP5406-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001241676.1 | |
IL-15 Protein | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the CTLL-2 cell proliferation assay. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
16168 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
IL15 | |
Recombinant Protein | |
E. coli | |
MNWIDVRYDL EKIESLIQSI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VAESGCKECE ELEEKTFTEF LQSFIRIVQM FINTS |