missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse IL-1ra/IL-1F3/IL1RN Protein

Product Code. p-200055688 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
1mg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418251 100 μg 100µg
18495002 1mg 1mg
18488371 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18418251 Supplier Novus Biologicals™ Supplier No. NBP235105100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

IL-1ra/IL-1F3/IL1RN protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting IL-1alpha- dependent proliferation of murine D10S cells is less than 50 ng/mL, corresponding to a specific activity of >2.0 x 104 IU/mg in the presence of 50 pg/mL rHuIL-1alpha.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q542W1, Q542W1, Q542W1
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 3557
Molecular Weight (g/mol) M.W. (Theoretical): 17.3 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of IL-1ra/IL-1F3/IL1RN as determined by LAL method.
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Gene Symbol IL1RN
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Research Category Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Purity or Quality Grade >97%pure by SDS-PAGE and HPLC
Protein IL-1ra/IL-1F3/IL1RN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.