missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse IL-36 gamma/IL-1F9 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.01 mg
1000 μg
Unit Size:
0.01mg
1000µg
Description
Fully biologically active when compared to standard. The ED50 as determined by inducing IL-6 secretion in murine NIH/3T3 cells is less than 10 ng/mL, corresponding to a specific activity of > 1.0 x 10^5 IU/mg.
Specifications
Specifications
| Accession Number | Q8R460, Q8R460, Q8R460 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in 1 M MOPS, 10 mM NaAC, pH7.6, 2 mM EDTA, 0.02 % Tween-20. |
| Gene ID (Entrez) | 56300 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 17.3 kDa |
| Quantity | 0.01 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of IL-36 gamma/IL-1F9 as determined by LAL method. |
| Gene Alias | IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1 RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma |
| Gene Symbol | IL36G |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction