missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL17 / IL17A Protein

Recombinant Protein

Brand:  enQuireBio™ QP5466-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956189

  • 4060.00 € / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_034682.1
IL17 / IL17A Protein
Research Use Only
IL17A
Recombinant Protein
E. coli
MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA
Greater than 95% as determined by SDS-PAGE.
16171
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 5 ng/mL in the Production of IL-6 by mouse 3T3 cells assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg.
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers