missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse KGF Protein

A cDNA sequence encoding the KGF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10734-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955008

  • 6790.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KGF Protein
Research Use Only
Recombinant Protein
E. coli
MCNDMSPEQT ATSVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNSYNIMEI RTVAVGIVAI KGVESEYYLA MNKEGKLYAK KECNEDCNFK ELILENHYNT YASAKWTHSG GEMFVALNQK GIPVKGKKTK KEQKTAHFLP MAIT
Greater than 95.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Mouse
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM Phosphate buffer pH 8 and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers