missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse KGF Protein
A cDNA sequence encoding the KGF was constructed and used to recombinantly synthesize the protein.
131.00 € - 6790.00 €
Specifications
Name | KGF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965008
|
enQuireBio™
QP10734-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945008
|
enQuireBio™
QP10734-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955008
|
enQuireBio™
QP10734-1MG |
1 mg |
6790.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
KGF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM Phosphate buffer pH 8 and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MCNDMSPEQT ATSVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNSYNIMEI RTVAVGIVAI KGVESEYYLA MNKEGKLYAK KECNEDCNFK ELILENHYNT YASAKWTHSG GEMFVALNQK GIPVKGKKTK KEQKTAHFLP MAIT | |
Greater than 95.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE. |