missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse LIGHT/TNFSF14 Protein

Product Code. 18608858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18608858 1000 μg 1000µg
18665727 500 μg 500µg
18622787 100 μg 100µg
18647356 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18608858 Supplier R&D Systems™ Supplier No. NBP2613321000ug
  • ONLINE EXCLUSIVE PRICE
    4404.48 €
    3115.35 € / 1000µg
    Save 1289.13 €
    29% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

LIGHT/TNFSF14 protein is fully biologically active when compared to standard. The ED50 as determined by a cytotoxicity assay using human HT-29 cells is less than 2 μg/mL, corresponding to a specific activity of > 500 IU/mg in the presence of murine anti-polyHistidine monoclonal antibody and rHuIFN-g.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH7.4.
Gene ID (Entrez) 8740
Molecular Weight (g/mol) MolecularWeight-theroretical: 18.4 kDa
Quantity 1000 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of LIGHT/TNFSF14 as determined by LAL method.
Gene Alias CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Gene Symbol TNFSF14
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in 100 mM HAC to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Apoptosis
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein LIGHT/TNFSF14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.