missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LY-96 / ESOP-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) LY-96 / ESOP-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6327-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001153183.1 | |
LY-96 / ESOP-1 Protein | |
Research Use Only | |
LY96 | |
Recombinant Protein | |
E. coli | |
EKQQWFCNSS DAIISYSYCD HLKFPISISS EPCIRLRGTN GFVHVEFIPR GNLKYLYFNL FISVNSIELP KRKEVLCHGH DDDYSFCRAL KGETVNTSIP FSFEGILFPK GHYRCVAEAI AGDTEEKLFC LNFTIIHRRD VN | |
Greater than 90% as determined by SDS-PAGE. |
17087 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |