missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LY-96 / ESOP-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) LY-96 / ESOP-1, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001153183.1 |
---|---|
Gene ID (Entrez) | 17087 |
Name | LY-96 / ESOP-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022500
|
enQuireBio™
QP6327-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042500
|
enQuireBio™
QP6327-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032500
|
enQuireBio™
QP6327-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052500
|
enQuireBio™
QP6327-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062500
|
enQuireBio™
QP6327-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072500
|
enQuireBio™
QP6327-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001153183.1 | |
LY-96 / ESOP-1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
17087 | |
Research Use Only | |
LY96 | |
Recombinant Protein | |
E. coli | |
EKQQWFCNSS DAIISYSYCD HLKFPISISS EPCIRLRGTN GFVHVEFIPR GNLKYLYFNL FISVNSIELP KRKEVLCHGH DDDYSFCRAL KGETVNTSIP FSFEGILFPK GHYRCVAEAI AGDTEEKLFC LNFTIIHRRD VN | |
Greater than 90% as determined by SDS-PAGE. |