missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LYVE1 Protein
A cDNA sequence encoding the LYVE1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12610-10ug
Additional Details : Weight : 0.01000kg
Specifications
LYVE1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MLQHTSLVLL LASIWTTRHP VQGADLVQDL SISTCRIMGV ALVGRNKNPQ MNFTEANEAC KMLGLTLASR DQVESAQKSG FETCSYGWVG EQFSVIPRIF SNPRCGKNGK GVLIWNAPSS QKFKAYCHNS SDTWVNSCIP EIVTTFYPVL DTQTPATEFS VSSSAYLASS PDSTTPVSAT TRAPPLTSMA RKTKKICITE VYTEPITMAT ETEAFVASGA AFKNEAAGFG GVPTALLVLA LLFFGAAAVL AVCYVKRYVK AFPFTTKNQQ KEMIETKVVK EEKADDVNAN EESKKTIKNP EEAKSPPKTT VRCLEAEV | |
Greater than 95.0% as determined by:(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
LYVE1 was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |