missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LYVE1 Protein
A cDNA sequence encoding the LYVE1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | LYVE1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940989
|
enQuireBio™
QP12610-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920989
|
enQuireBio™
QP12610-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930989
|
enQuireBio™
QP12610-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
LYVE1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
LYVE1 was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MLQHTSLVLL LASIWTTRHP VQGADLVQDL SISTCRIMGV ALVGRNKNPQ MNFTEANEAC KMLGLTLASR DQVESAQKSG FETCSYGWVG EQFSVIPRIF SNPRCGKNGK GVLIWNAPSS QKFKAYCHNS SDTWVNSCIP EIVTTFYPVL DTQTPATEFS VSSSAYLASS PDSTTPVSAT TRAPPLTSMA RKTKKICITE VYTEPITMAT ETEAFVASGA AFKNEAAGFG GVPTALLVLA LLFFGAAAVL AVCYVKRYVK AFPFTTKNQQ KEMIETKVVK EEKADDVNAN EESKKTIKNP EEAKSPPKTT VRCLEAEV | |
Greater than 95.0% as determined by:(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE. |