missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCP 1 Protein
A cDNA sequence encoding the MCP 1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10781-2ug
Additional Details : Weight : 0.01000kg
Specifications
20296 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity was determined by calculating its ability to chemoattract Balb/C mouse spleen MNCs at 1.0-20.0 ng/ml. | |
Mouse | |
Untagged | |
The protein was lyophilized with no additives. |
MCP 1 Protein | |
Research Use Only | |
Ccl2 | |
Recombinant Protein | |
E. coli | |
MQVPVMLLGL LFTVAGWSIH VLAQPDAVNA PLTCCYSFTS KMIPMSRLES YKRITSSRCP KEAVVFVTKL KREVCADPKK EWVQTYIKNL DRNQMRSEPT TLFKTASALR SSAPLNVKLT RKSEANASTT FSTTTSSTSV GVTSVTVN | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |