missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCP 1 Protein
A cDNA sequence encoding the MCP 1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 20296 |
---|---|
Name | MCP 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Ccl2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995148
|
enQuireBio™
QP10781-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975148
|
enQuireBio™
QP10781-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985148
|
enQuireBio™
QP10781-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
20296 | |
Research Use Only | |
Ccl2 | |
Recombinant Protein | |
E. coli | |
MQVPVMLLGL LFTVAGWSIH VLAQPDAVNA PLTCCYSFTS KMIPMSRLES YKRITSSRCP KEAVVFVTKL KREVCADPKK EWVQTYIKNL DRNQMRSEPT TLFKTASALR SSAPLNVKLT RKSEANASTT FSTTTSSTSV GVTSVTVN | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
MCP 1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity was determined by calculating its ability to chemoattract Balb/C mouse spleen MNCs at 1.0-20.0 ng/ml. | |
Mouse | |
Untagged | |
The protein was lyophilized with no additives. |