missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Osteocalcin Protein
A DNA sequence encoding the Mus musculus (Mouse) Osteocalcin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5716-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031567.1 | |
Osteocalcin Protein | |
Research Use Only | |
Bglap | |
Recombinant Protein | |
E. coli | |
YLGASVPSPD PLEPTREQCE LNPACDELSD QYGLKTAYKR IYGITI | |
Greater than 90% as determined by SDS-PAGE. |
12096 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |