missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Osteocalcin Protein
A DNA sequence encoding the Mus musculus (Mouse) Osteocalcin, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_031567.1 |
---|---|
Gene ID (Entrez) | 12096 |
Name | Osteocalcin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927229
|
enQuireBio™
QP5716-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947229
|
enQuireBio™
QP5716-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937229
|
enQuireBio™
QP5716-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957229
|
enQuireBio™
QP5716-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15967229
|
enQuireBio™
QP5716-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15977229
|
enQuireBio™
QP5716-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031567.1 | |
Osteocalcin Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
12096 | |
Research Use Only | |
Bglap | |
Recombinant Protein | |
E. coli | |
YLGASVPSPD PLEPTREQCE LNPACDELSD QYGLKTAYKR IYGITI | |
Greater than 90% as determined by SDS-PAGE. |