missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Sushi repeat-containing protein SRPX2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Sushi repeat-containing protein SRPX2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7826-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031031

  • 3260.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001077364.2
Sushi repeat-containing protein SRPX2 Protein
Research Use Only
Srpx2
Recombinant Protein
E. coli
WYAGSGYSPD ESYNEVYAEE VPAARARALD YRVPRWCYTL NIQDGEATCY SPRGGNYHSS LGTRCELSCD RGFRLIGRKS VQCLPSRRWS GTAYCRQIRC HTLPFITSGT YTCTNGMLLD SRCDYSCSSG YHLEGDRSRI CMEDGRWSGG EPVCVDIDPP KIRCPHSREK MAEPEKLTAR VYWDPPLVKD SADGTITRVT LRGPEPGSHF PEGEHVIRYT AYDRAYNRAS CKFIVKVQVR RCPILKPPQH GYLTCSSAGD NYGAICEYHC DGGYERQGTP SRVCQSSRQW SGTPPVCTPM KINVNVNSAA GLLDQFYEKQ RLLIVSAPDP SNRYYKMQIS MLQQSTCGLD LRHVTIIELV GQPPQEVGRI REQQLSAGII EELRQFQRLT RSYFNMVLID KQGIDRERYM EPVTPEEIFT FIDDYLLSNE ELARRVEQRD LCE
Greater than 80% as determined by SDS-PAGE.
68792
1 mg
Not determined.
Full Length of Mature Protein
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers