missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TNFSF14 Protein

A cDNA sequence encoding the TNFSF14 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10890-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15975478

  • 3915.00 € / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

50930
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by a cytotoxicity assay using human HT-29 cells is less than 2microg/ml, corresponding to a specific activity of > 500 IU/mg in the presence of murine anti-polyHistidine monoclonal antibody and rHuIFN-g.
Mouse
Untagged
TNFSF14 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4.
TNFSF14 Protein
Research Use Only
Tnfsf14
Recombinant Protein
E. coli
DGGKGSWEKL IQDQRSHQAN PAAHLTGANA SLIGIGGPLL WETRLGLAFL RGLTYHDGAL VTMEPGYYYV YSKVQLSGVG CPQGLANGLP ITHGLYKRTS RYPKELELLV SRRSPCGRAN SSRVWWDSSF LGGVVHLEAG EEVVVRVPGN RLVRPRDGTR SYFGAFMV
Greater than 96.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers