missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TNFSF14 Protein
A cDNA sequence encoding the TNFSF14 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 50930 |
---|---|
Name | TNFSF14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Tnfsf14 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995478
|
enQuireBio™
QP10890-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985478
|
enQuireBio™
QP10890-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975478
|
enQuireBio™
QP10890-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
50930 | |
Research Use Only | |
Tnfsf14 | |
Recombinant Protein | |
E. coli | |
DGGKGSWEKL IQDQRSHQAN PAAHLTGANA SLIGIGGPLL WETRLGLAFL RGLTYHDGAL VTMEPGYYYV YSKVQLSGVG CPQGLANGLP ITHGLYKRTS RYPKELELLV SRRSPCGRAN SSRVWWDSSF LGGVVHLEAG EEVVVRVPGN RLVRPRDGTR SYFGAFMV | |
Greater than 96.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
TNFSF14 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cytotoxicity assay using human HT-29 cells is less than 2microg/ml, corresponding to a specific activity of > 500 IU/mg in the presence of murine anti-polyHistidine monoclonal antibody and rHuIFN-g. | |
Mouse | |
Untagged | |
TNFSF14 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4. |