missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TNFSF14 Protein
A cDNA sequence encoding the TNFSF14 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10890-20ug
Additional Details : Weight : 0.01000kg
Specifications
50930 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cytotoxicity assay using human HT-29 cells is less than 2microg/ml, corresponding to a specific activity of > 500 IU/mg in the presence of murine anti-polyHistidine monoclonal antibody and rHuIFN-g. | |
Mouse | |
Untagged | |
TNFSF14 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4. |
TNFSF14 Protein | |
Research Use Only | |
Tnfsf14 | |
Recombinant Protein | |
E. coli | |
DGGKGSWEKL IQDQRSHQAN PAAHLTGANA SLIGIGGPLL WETRLGLAFL RGLTYHDGAL VTMEPGYYYV YSKVQLSGVG CPQGLANGLP ITHGLYKRTS RYPKELELLV SRRSPCGRAN SSRVWWDSSF LGGVVHLEAG EEVVVRVPGN RLVRPRDGTR SYFGAFMV | |
Greater than 96.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |