Learn More
enQuireBio™ Recombinant other Avidin Biotin Protein
Recombinant Protein
Brand:  enQuireBio™ QP10526-100mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15964138
  • 1440.00 € / 100mg
Estimated Shipment: 04-07-2024
to see stock.
Specifications
396260
100 mg
< 1.0 EU per ug protein as determined by the LAL method.
Binding activity of Avidin-Biotin is 10-15units/mg protein. One unit of Avidin activity is defined as the amount of protein which will bind 1microg d-biotin.
Other
MVHATSPLLL LLLLSLALVA PGLSARKCSL TGKWTNDLGS NMTIGAVNSR GEFTGTYITA VTATSNEIKE SPLHGTQNTI NKRTQPTFGF TVNWKFSEST TVFTGQCFID RNGKEVLKTM WLLRSSVNDI GDDWKATRVG INIFTRLRTQ KE
Avidin Biotin Protein
Research Use Only
AVD
Recombinant Protein
Untagged
Salt-free lyophilized powder with no preservative added.