missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Avidin Biotin Protein
Recombinant Protein
131.00 € - 1440.00 €
Specifications
Gene ID (Entrez) | 396260 |
---|---|
Name | Avidin Biotin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | AVD |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15984138
|
enQuireBio™
QP10526-2MG |
2 mg |
131.00 €
2mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974138
|
enQuireBio™
QP10526-10MG |
10 mg |
203.00 €
10mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964138
|
enQuireBio™
QP10526-100MG |
100 mg |
1440.00 €
100mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
396260 | |
Research Use Only | |
AVD | |
Recombinant Protein | |
Untagged | |
Salt-free lyophilized powder with no preservative added. |
Avidin Biotin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Binding activity of Avidin-Biotin is 10-15units/mg protein. One unit of Avidin activity is defined as the amount of protein which will bind 1microg d-biotin. | |
Other | |
MVHATSPLLL LLLLSLALVA PGLSARKCSL TGKWTNDLGS NMTIGAVNSR GEFTGTYITA VTATSNEIKE SPLHGTQNTI NKRTQPTFGF TVNWKFSEST TVFTGQCFID RNGKEVLKTM WLLRSSVNDI GDDWKATRVG INIFTRLRTQ KE |