missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia p41 Protein
A cDNA sequence encoding the Borrelia p41 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11192-HIS-500ug
Additional Details : Weight : 0.01000kg
Specifications
Borrelia p41 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM glycine, pH 9.6 and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >90% pure as determined by SDS-PAGE. |