missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia p41 Protein
A cDNA sequence encoding the Borrelia p41 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | Borrelia p41 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976428
|
enQuireBio™
QP11192-HIS-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996428
|
enQuireBio™
QP11192-HIS-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986428
|
enQuireBio™
QP11192-HIS-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Borrelia p41 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >90% pure as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM glycine, pH 9.6 and 50% glycerol. |