missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Ganirelix Protein
A cDNA sequence encoding the Ganirelix was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11930-500ug
Additional Details : Weight : 0.01000kg
Specifications
Ganirelix Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Ganirelix hormone (0.5 mg/ml) contains 0.1 mg acetic acid and 23.5 mg mannitol pH 5. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |