missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Ganirelix Protein
A cDNA sequence encoding the Ganirelix was constructed and used to recombinantly synthesize the protein.
228.00 € - 955.00 €
Specifications
Name | Ganirelix Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15988828
|
enQuireBio™
QP11930-50UG |
50 μg |
228.00 €
50µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968828
|
enQuireBio™
QP11930-150UG |
150 μg |
373.00 €
150µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978828
|
enQuireBio™
QP11930-500UG |
500 μg |
955.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Ganirelix Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Ganirelix hormone (0.5 mg/ml) contains 0.1 mg acetic acid and 23.5 mg mannitol pH 5. |