missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Internalin-A Protein
A cDNA sequence encoding the Internalin-A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12452-20ug
Additional Details : Weight : 0.01000kg
Specifications
Internalin-A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MANENNFLEI MSETKEIKIV NIPDNNLKKV LNKSLNKSEN SDLTVKDLES IEYLYGIAEN ISNIEGLEYC KNLKILSLQN NDNSKKENFN TITDLSPLKY LKNLVVLDLR NNKISDLSPL ENLTNLESLR LSGNNISNIS PLNKLESLTT LTLSYNEITD ISTISNLKNL THLALYNNKI EDISSLKENT KLQNLSLGFN KIKDISVLSN LKNLYDLSLE ENNIKSIKSL SNLHKLSNIN LKNNKIEDIS PLETCNDFFE KLILDGNRIS DISSLSKKEI IVCSINNQTI TIKDWKVSDN NSFEIKLPND NLDIKIDNIS ENGTFIDNSI KWTNISPKSK LSFEFKQDID NDIHSIFYSG TVRFKRLEHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE and HPLC. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Internalin-A is lyophilized from a 0.2microm filtered concentrated solution in 20mM PB pH 7.4 and 300mM NaCl. |