missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Internalin-A Protein
A cDNA sequence encoding the Internalin-A was constructed and used to recombinantly synthesize the protein.
252.00 € - 3915.00 €
Specifications
Name | Internalin-A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15900469
|
enQuireBio™
QP12452-20UG |
20 μg |
252.00 €
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980459
|
enQuireBio™
QP12452-100UG |
100 μg |
520.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15990459
|
enQuireBio™
QP12452-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Internalin-A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Internalin-A is lyophilized from a 0.2microm filtered concentrated solution in 20mM PB pH 7.4 and 300mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MANENNFLEI MSETKEIKIV NIPDNNLKKV LNKSLNKSEN SDLTVKDLES IEYLYGIAEN ISNIEGLEYC KNLKILSLQN NDNSKKENFN TITDLSPLKY LKNLVVLDLR NNKISDLSPL ENLTNLESLR LSGNNISNIS PLNKLESLTT LTLSYNEITD ISTISNLKNL THLALYNNKI EDISSLKENT KLQNLSLGFN KIKDISVLSN LKNLYDLSLE ENNIKSIKSL SNLHKLSNIN LKNNKIEDIS PLETCNDFFE KLILDGNRIS DISSLSKKEI IVCSINNQTI TIKDWKVSDN NSFEIKLPND NLDIKIDNIS ENGTFIDNSI KWTNISPKSK LSFEFKQDID NDIHSIFYSG TVRFKRLEHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE and HPLC. |