missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MSRA Protein
A cDNA sequence encoding the MSRA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12742-5ug
Additional Details : Weight : 0.01000kg
Specifications
MSRA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSLFDKKHLV SPADALPGRN TPMPVATLHA VNGHSMTNVP DGMEIAIFAM GCFWGVERLF WQLPGVYSTA AGYTGGYTPN PTYREVCSGD TGHAEAVRIV YDPSVISYEQ LLQVFWENHD PAQGMRQGND HGTQYRSAIY PLTPEQDAAA RASLERFQAA MLAADDDRHI TTEIANATPF YYAEDDHQQY LHKNPYGYCG IGGIGVCLPP EA | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
MSRA protein solution (0.5 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |