missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PCC Protein
A cDNA sequence encoding the PCC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12973-10IU
Additional Details : Weight : 0.01000kg
Specifications
PCC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized Powder from 0.1M sodium chloride and 10mM sodium citrate buffer. |
10 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |