missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Thymosin ?4 Protein

A cDNA sequence encoding the Thymosin ?4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13746-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934569

  • 1320.00 € / 1mg
Estimated Shipment: 12-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Thymosin ?4 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Lyophilized from a 0.2um filtered concentrated solution in 1xPBS, pH 7.4.
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses.
Research Use Only
Recombinant Protein
E. coli
SDKPDMAEIE KFDKSKLKKT ETQEKNPLPS KETIEQEKQA GES
Videos
SDS
Documents
Certificates
Special Offers

Special Offers