missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Plant Arachis hypogaea 8.0101 Strep (N-Term) Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Lyophilized from a 0.22 um filtered solution in Tris buffer (100 mM Tris, 150 mM NaCl, pH 8) |
| Molecular Weight (g/mol) | M.W. Theoretical: 18.3 kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -20 to -70C. Avoid freeze-thaw cycles. |
| Gene Alias | rAra h 8 |
| Reconstitution | Reconstitute with sterile deionized water with or without azide. |
| Purity or Quality Grade | >95%, by HPLC |
| Protein | Arachis hypogaea 8.0101 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction