missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Plant Phleum pratense 2.0101 Strep (N-Term) Protein

Product Code. 18324725 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18324725

Brand: Novus Biologicals™ NBP306726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution in Tris buffer (100 mM Tris, 150 mM NaCl, pH 8)
Molecular Weight (g/mol) M.W. Theoretical: 12.3 kDa
Quantity 0.1 mg
Storage Requirements Store at -20 to -70C. Avoid freeze-thaw cycles.
Gene Alias rPhl p 2
Reconstitution Reconstitute with sterile deionized water with or without azide.
Purity or Quality Grade >95%, by HPLC
Protein Phleum pratense 2.0101
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.