missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Trypsin Protein
A cDNA sequence encoding the Trypsin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10897-1mg
Additional Details : Weight : 0.01000kg
Specifications
Trypsin Protein | |
Research Use Only | |
Recombinant Protein | |
P. pastoris | |
VGGYTCAANSIPYQVSLNSGSHFCGGSLINSQWVVSAAHCYKSRIQVRLGEHNI DVLEGNEQFINAAKIITHPNFNGNTLDNDIMLIKLSSPATLNSRVATVSLPRSCA AAGTECLISGWGNTKSSGSSYPSLLQCLKAPVLSDSSCKSSYPGQITGNMICVGF LEGGKDSCQGDSGGPVVCNGQLQGIVSWGYGCAQKNKPGVYTKVCNYVNWI QQTIAAN |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Porcine | |
Untagged | |
The Porcine Trypsin (2.98 mg/ml) is formulated with 1mM HCl and 20mM CaCl2, pH 3. |