missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Protein L Type 1 His (N-Term) Cys (C-Term) Protein
Shop All Bio Techne ProductsDescription
Protein L is a genetically engineered protein containing 5 IgG-binding regions of protein L. Cell wall binding region, cell membrane binding region and albumin binding region have been removed from the recombinant Protein L to ensure the maximum specific IgG binding. Protein L binds to human, mouse, rat and pig IgG.
Specifications
Specifications
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from additive free solution. |
| Molecular Weight (g/mol) | 41.6 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of Protein L Type 1 as determined by LAL method. |
| Reconstitution | Reconstitute with distilled water or saline. |
| Research Category | Biologically Active Proteins |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | Protein L Type 1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?