missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit VCAM1 Protein
A cDNA sequence encoding the VCAM1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13929-10ug
Additional Details : Weight : 0.01000kg
Specifications
100008901 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
AFKIETFPESRSLAQIGDSVSLTCTTTGCASPTFSWRTQIDSPLNGKVRSEGTTSTLTMDPVSFENE HSYLCTATCESKKLEKGIQVEIYSFPKDPEIHLSGPLEVGEPITVKCLVPDVYPFDRLEVDLLKGDYLM KKQDFLEDMDRKSLETKSLEVTFIPVIEDIGKLIVCRAKLHIDEIDSEPKERETTKELQVYISPKNTVIS VNPSTRLQEGGSVTMTCSSEGLPVPEIFWSKKQDNGNLQRLSGNATLTLIAMRMEDSGIYVCEG VNQIGKSRKEVELIVQEKPFTVEISPGPRIAAQIGDPVVLTCSVRGCETPSFSWRTQIDSPLNGQVT SEGTKSLLTLSPVSFENEHSYLCTVTCGHKKLEKGIQVELYSFPRDPEIELSGPPVNGRPVTVSCKVP NVYPFDXLEIELLKGETMMKNKEFLEEEDKKSLETKSLEMTFIPTMEDTGKVLVCQAKLHIDEMEF EPKQRQSTQPLFVNVAPRDIAVWVSPSSIVEEGRSVNMTCSSYGLPAPKILWSRQLKNGDLQPLS ENTTLALISTKLEDSGIYVCEGINLAGKSRKEVELVIQVAPKDIQLTAFPSKSVKEGDTVIISCTCGNV PETWIILKKKAETGDTVLKSIDGAYTIRKAQLEDAGVYECESKNEVGSQLRSITLDVKGRENSKDYF SPE | |
Greater than 90% as determined by SDS-PAGE. |
VCAM1 Protein | |
Research Use Only | |
VCAM-1 | |
Rabbit | |
His | |
The VCAM1 solution (1 mg/ml) contains 50mM Tris, 300mM NaCl, 10% Glycerol, pH 7.5. |