missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rainbow Trout PRL R Protein
A cDNA sequence encoding the PRL R was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10844-5ug
Additional Details : Weight : 0.01000kg
Specifications
PRL R Protein | |
Research Use Only | |
Activity is determined by the dose-dependant inhibition of Prolactin-stimuled proliferation of Nb2 cells and by high affinity binding of oPLR and other lactogenic hormones. | |
Rainbow Trout | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MMWRDVGVTL ILSLILPVVA QGARHTPPGK PKLTSCRSPD KETFTCWWEA GSDGGRPTTY SLFYHKENSE TVYECPDYHT AGANSCFFNK NETSIWVNYN ITVVATNALG NNFSDPVDVD VVYIVQPHTP ENVTVGVLED EDGPFLRVSW EPPRKADTRS GWITLIYELR VKLEEAEEWE EHRAGQQKMF NNFSLHSGGV YMVQVRCKPD HGFWSEWSTA SYVKVPDYIP RERSMWILIA VFSAFIFLIL TWIITLNRSS VKHCLLPPVP GPKIKGFDQQ LLKKGKPEEV FNSLVVQSFP PRSTDYEDLL VEYLEVYVNE KQELMPEGKD LRDVGCLESK SQSDNDSGRG SCDSHTLLME KCGGVESKDE SSYEEPELLQ GQAGSWERLE RGDSQVVDTP DSSDARVKTW PLVFSPSIHG SSDNHHYGTL EMPKQHSTSE ITYSVPDHLF PTSTSFPHHH HPEYRDSLGE YSEYRHSRSP STGVLYHWEA QHHPQAHSNF NIRNLERQKE ATGFGLPPSR SMEYVEVQKV NQNQLVLKPL SSHGRGPSQV QFGRAVVDYS KVNGVNNDNV LLLQRQREMA EAGQYCDYRE KEGAEAERYP TQQQGKTTKD GPVATQLQES TCLTTSGYVD TVPTMPTF | |
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |