missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rainbow Trout PRL R Protein
A cDNA sequence encoding the PRL R was constructed and used to recombinantly synthesize the protein.
131.00 € - 4880.00 €
Specifications
Name | PRL R Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Activity is determined by the dose-dependant inhibition of Prolactin-stimuled proliferation of Nb2 cells and by high affinity binding of oPLR and other lactogenic hormones. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985338
|
enQuireBio™
QP10844-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975338
|
enQuireBio™
QP10844-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965338
|
enQuireBio™
QP10844-1MG |
1 mg |
4880.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PRL R Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MMWRDVGVTL ILSLILPVVA QGARHTPPGK PKLTSCRSPD KETFTCWWEA GSDGGRPTTY SLFYHKENSE TVYECPDYHT AGANSCFFNK NETSIWVNYN ITVVATNALG NNFSDPVDVD VVYIVQPHTP ENVTVGVLED EDGPFLRVSW EPPRKADTRS GWITLIYELR VKLEEAEEWE EHRAGQQKMF NNFSLHSGGV YMVQVRCKPD HGFWSEWSTA SYVKVPDYIP RERSMWILIA VFSAFIFLIL TWIITLNRSS VKHCLLPPVP GPKIKGFDQQ LLKKGKPEEV FNSLVVQSFP PRSTDYEDLL VEYLEVYVNE KQELMPEGKD LRDVGCLESK SQSDNDSGRG SCDSHTLLME KCGGVESKDE SSYEEPELLQ GQAGSWERLE RGDSQVVDTP DSSDARVKTW PLVFSPSIHG SSDNHHYGTL EMPKQHSTSE ITYSVPDHLF PTSTSFPHHH HPEYRDSLGE YSEYRHSRSP STGVLYHWEA QHHPQAHSNF NIRNLERQKE ATGFGLPPSR SMEYVEVQKV NQNQLVLKPL SSHGRGPSQV QFGRAVVDYS KVNGVNNDNV LLLQRQREMA EAGQYCDYRE KEGAEAERYP TQQQGKTTKD GPVATQLQES TCLTTSGYVD TVPTMPTF | |
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Activity is determined by the dose-dependant inhibition of Prolactin-stimuled proliferation of Nb2 cells and by high affinity binding of oPLR and other lactogenic hormones. | |
Rainbow Trout | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |