missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat CCL24/Eotaxin-2/MPIF-2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
Eotaxin 2 Protein is fully biologically active when compared to standard. The biologically active determined by a chemotaxis bioassay using human peripheral blood eosinophils is in a concentration of 50-250 ng/mL.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 2 x PBS, pH 7.4. |
| Gene ID (Entrez) | 6369 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 10.5 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of CCL24/Eotaxin-2/MPIF-2 AS determined by LAL method. |
| Gene Alias | CCL24, chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, Eotaxin-2/CCL24, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2 Small-inducible cytokine A24, SCYA24CK-beta-6 |
| Gene Symbol | CCL24 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction